Learn More
Gas Arc™ Inline Flashback Arrestor
POA
Product Code | Brand | For Use With (Application) | Pressure | Connection Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Pressure | Connection Type | Price | Quantity & Availability | |||
12960820
![]() |
Gas Arc™
FBA07048 |
Fuel Gas | 5 bar (inlet)/5 bar (outlet) | 3/8 BSP LH Female (inlet); 3/8 BSP LH Male (outlet) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
12950820
![]() |
Gas Arc™
FBA07051 |
Oxygen | 20 bar (inlet)/20 bar (outlet) | 3/8 BSP RH Female (inlet); 3/8 BSP RH Male (outlet) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
Specifications
Inline Flashback Arrestor |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Gas Arc™ Inline Flashback Arrestor