Learn More
Memmert™ Flexible Pt100 Temperature Sensor for UF110plus
Flexible temperature sensor for positioning in chamber or in load.
Brand:  Memmert™ UF110plusH8
Product Code. 17528985
  • 2605.00 SEK / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Description
  • Positioned flexibly in chamber or load
  • For local temperature measurement (up to 1 additional sensor is possible)
  • The measured temperature will be indicated on the display, recorded in the integral data store, and can be documented via the AtmoCONTROL software
  • For Memmert Ovens
Specifications
Specifications
UF110plus Oven
Temperature Sensor
Product Title
Memmert™ Flexible Pt100 Temperature Sensor for UF110plus > For Use With: UF110plus