missing translation for 'onlineSavingsMsg'
Learn More

Systec™ Floor Model, Pass-Through Autoclave, HX Series 2D

Systec™ Floor Model, Pass-Through Autoclave, HX Series 2D are designed to be built into a wall for clean room operations.

Brand:  Systec™ 2235

 View more versions of this product

Product Code. 12572767

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Highest requirements with regard to safe, accurate, reproducible and validatable sterilization processes.
  • For use under the most stringent clean room and safety conditions: For biological safety laboratories, e.g., fitting as a sterilization and pass-through lock for protecting the external environment, for clean rooms in laboratories and production facilities as a sterilization and pass-through lock separating sterile and non-sterile areas.
  • Fully automatic, micro-processor controlled.
  • Can be used for all laboratory applications, even for sophisticated sterilization processes.
  • All additional optional accessories can be fitted to obtain validatable processes.
  • Standard features:
    • Integrated, separate steam generator
    • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
    • Touch-Screen control
    • Number of sterilization programs: up to 100
    • Code-secured access rights for changing parameters and further safety-relevant intervention
    • Internal memory for storing up to 500 sterilization cycles
    • Timer for starting programs
    • Autofill: automatic demineralized water feed for steam generation
    • Flexible PT-100 temperature sensor
    • Additional temperature sensor in condense exhaust
    • Temperature holding function for liquids after program finish
    • Special program for Durham tubes
    • Calculation of F0 value
    • Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
    • Water-cooled steam exhaust, thermostatically controlled
    • Programmable automatic door-opening on completion of program
    • RS-232 and RS-485 interfaces for external data transmission (network-compatible)
  • Available options include temperature, pressure, cooling, vacuum, drying, exhaust, and documentation options.
Specifications

Specifications

370 L (Total), 320 L (Nominal)
740 mm
Double-Door/Pass-Through Autoclave
4 bar
750 mm
HX-320 2D
140°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers