Learn More
STEMCELL Technologies Fluoxetine (Hydrochloride)
Serotonin reuptake inhibitor
POA
Specifications
CAS 56296-78-7
Molecular Formula C17H18F3NO · HCl
Purity ≥98%
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17100182
Encompass_Preferred
View Documents
STEMCELL Technologies
73142
100 mg
POA
100mg
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17110182
Encompass_Preferred
View Documents
STEMCELL Technologies
73144
500 mg
POA
500mg
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • Fluoxetine is a selective serotonin reuptake inhibitor, displaying a distinct preference for the serotonin transporter (Kd = 0.81 nM) over both the norepinephrine and dopamine transporters (Kd values of 240 and 3600 nM, respectively; Tatsumi et al.) This product is supplied as a hydrochloride salt of the molecule
  • Induces proliferation of human embryonic stem cell-derived neural precursors, and enhances their neuronal differentiation (Chang et al. 2010)
  • Induces proliferation and inhibits differentiation of human hypothalamic neuroprogenitor cells (Sousa-Ferreira et al.)
  • Stimulates proliferation of mouse fetal neural stem cells (Chang et al. 2012)
  • Stimulates in vivo proliferation of amplifying neural progenitor cells in adult mouse brain (Encinas et al.)
Specifications
Specifications
56296-78-7
C17H18F3NO · HCl
≥98%
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.