missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FMR1 Mouse anti-Human, Mouse, Rat, Clone: 2B5B5, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | FMR1 |
---|---|
Clone | 2B5B5 |
Concentration | 1 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16868525
|
Proteintech
66548-1-IG-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16858525
|
Proteintech
66548-1-IG-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Members of the Rho.Specifications
FMR1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FMR1, FMRP, fragile X mental retardation 1, FRAXA, POF, POF1, Protein FMR 1 | |
FMR1 | |
IgG2a | |
Protein A | |
Antibody |
2B5B5 | |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Rat, Mouse, Human | |
P35922, Q06787, Q80WE1 | |
14265, 2332, 24948 | |
FMR1 Fusion Protein Ag4697 | |
Primary | |
-20°C | |
FMR1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FMR1 Mouse anti-Human, Mouse, Rat, Clone: 2B5B5, Proteintech