missing translation for 'onlineSavingsMsg'
Learn More

FOLH1B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

4460.00 SEK

Specifications

Antigen FOLH1B
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18332275
View Documents
Bio-Techne
NBP3-09721-100UL
100 μg
4460.00 SEK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FOLH1B Polyclonal specifically detects FOLH1B in Human samples. It is validated for Western Blot.
Specifications

Specifications

FOLH1B
Western Blot
Unconjugated
Rabbit
Human
219595
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
The immunogen is a synthetic peptide directed towards the middle region of Human FOLH1B (NP_710163). Peptide sequence ELKSPDEGFEGKSLYESWTKKSPSPEFSGMPRISKLGSGNDFEVFFQRLG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.