Learn More
Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 023277.QA
Product Code. 15461127
  • 3620.00 SEK / Each
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Measures 0 to 100ppm HCHO in aqueous solution
Formaldehyde test strip
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX04J019 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number023277.QA
Lot NumberP27J046 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number023277.QA
Lot NumberX27I003 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog Number023277.QA
Lot NumberQ22I061 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog Number023277.QA
Product Title
Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals > 1kit