missing translation for 'onlineSavingsMsg'
Learn More

Formamidine acetate, 99%, Thermo Scientific Chemicals

618.17 SEK - 2201.22 SEK

Chemical Identifiers

CAS 3473-63-0
Molecular Formula CH4N2·C2H4O2
Molecular Weight (g/mol) 104.11
MDL Number MFCD00012866
InChI Key XPOLVIIHTDKJRY-UHFFFAOYSA-N
Synonym formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
PubChem CID 160693
IUPAC Name acetic acid;methanimidamide
SMILES CC(=O)O.C(=N)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10734611
View Documents
Thermo Scientific Acros
119711000
100 g Plastic bottle
618.17 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
10645922
View Documents
Thermo Scientific Acros
119715000
500 g Plastic bottle
2201.22 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3473-63-0
104.11
XPOLVIIHTDKJRY-UHFFFAOYSA-N
160693
CC(=O)O.C(=N)N
CH4N2·C2H4O2
MFCD00012866
formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
acetic acid;methanimidamide

Specifications

3473-63-0
Colorless to Yellow
Authentic
CH4N2·C2H4O2
MFCD00012866
1.124
Solubility in water: 832g/L (21°C)
CC(=O)O.C(=N)N
104.11
104.11
Liquid
157°C to 161°C
1.1240g/mL
99%
HC(=NH)NH2·CH3CO2H
01,403; 02,202; 03,147
formamidine acetate, methanimidamide, monoacetate, formamidinium acetate, acetic acid; methanimidamide, formamidine acetate salt, formamidine, monoacetate, formimidamide acetate, imidoformamide acetate, acetic acid; formamidine, methanimidamide monoacetate
XPOLVIIHTDKJRY-UHFFFAOYSA-N
acetic acid;methanimidamide
160693
99%
Formamidine acetate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 222-442-5

missing translation for 'tsca' : TSCA

Product Identifier
  • Formamidine acetate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.