missing translation for 'onlineSavingsMsg'
Learn More
Honeywell Safety Products™ Foundry Lined Glove Product Code.: 12691013

Honeywell Safety Products™ Foundry Lined Glove

Product Code. 12691013
Size: 10
Click to view available options
Size: 10
This item is not returnable. View return policy

Product Code. 12691013

Brand: Honeywell Safety Products™ 201284710

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Water-repellent and heat-resistant, this glove is constructed using split leather, grain leather, and non-flammable foam. Honeywell Safety Products™ Foundry Lined Glove is also designed with a protective cuff and stitched with sturdy, heat-resistant Kevlar™ thread.

  • Water-repellent and heat-resistant split cowhide leather construction suitable for welding.
  • Excellent resistance to contact and radiant heat.
  • Gunn cut, sewn-on thumb, gauntlet type design.
  • Cotton/viscose hand liner for improved touch.
  • Non-flammable foam lining laminated onto heat-resistant jersey.
  • U.S. assembly.
  • Winged thumb and construction materials provide flexibility.
  • Kevlar™ thread assembly ensures sturdy seams and better thermal insulation.
  • Grain leather cuff protects a large part of the forearm.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Thermical Hazards, EN 12477-2002 Type B Welding hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Attestation Number 0075/797/162/02/07/004
Classification Dexterity: 4
Cuff Material Grain Leather
Cut Style Gunn
Cut Resistance Rating 1
Material Cowhide Leather, Grain Leather, Cotton, Dupont™ Kevlar™ Aramid Fibers
Product Type Welding Gloves (Type B)
Size 10
Style Gauntlet
Thickness (Metric) 1.3/1.4 mm
For Use With (Application) Automotive, Foundry, Iron, Steel, Metal, Ship Building, Transportation, Utility and Welding industries.
Liner Material Cotton, Heat-resistant Foam
Lining Lined
Puncture Rating 3
Quantity 1 Pair/Pk.
Resistant To Heat, water
Tear Rating 3
Length (Metric) 38.5
Length (Metric) Cuff 18
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell Safety Products™ Foundry Lined Glove >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.