missing translation for 'onlineSavingsMsg'
Learn More

FoxP1 Mouse anti-Human, Clone: rFOXP1/6902, Novus Biologicals™

Mouse Monoclonal Antibody

2470.00 SEK - 4910.00 SEK

Specifications

Antigen FoxP1
Clone rFOXP1/6902
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18397034
View Documents
Bio-Techne
NBP3-13927-20UG
20 μg
2470.00 SEK
20µg
Please to purchase this item. Need a web account? Register with us today!
18332805
View Documents
Bio-Techne
NBP3-13927-100UG
100 μg
4910.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FoxP1 Monoclonal antibody specifically detects FoxP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

FoxP1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4
Recombinant fragment (around aa350-450) of human FoxP1 protein (exact sequence is proprietary) (Uniprot: Q9H334)
Primary
Store at 4°C.
rFOXP1/6902
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
Q9H334
27086
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.