Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 3 in/76 mm
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand: Eppendorf™ 6001000310
This item is not returnable.
View return policy
Description
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 3 in/76 mm, side access, stainless steel- Rack for 76 mm/3.0 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 141 x 141 x 726 (5.6 x 5.6 x 28.6 in)- Max. box number per rack: 9 (9 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 40 racks)- For ULT freezer: Innova C580 (max. 288 boxes), Innova C760 (max. 360 boxes), CryoCube™ FC660 series (max. 360 boxes)Specifications
Freezer Rack |