Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 4 in/102 mm
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Brand:  Eppendorf™ 6001062410
Product Code. 15793265
  • 1932.70 SEK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Freezer Rack: Innova™ U101, CryoCube™ F101h, 4 in/102 mm, drawer, stainless steel- Rack for 102 mm/4 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 308 x 145 x 216 (12.1 x 5.7 x 8.5 in)- Max. box number per rack: 4 (2 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 24 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Freezer Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products