Learn More
Frizzled 9 Mouse anti-Human, Unconjugated, Clone: 1G9G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67023-1-IG-20UL
This item is not returnable.
View return policy
Description
The Frizzled 9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Frizzled 9. This antibody recognizes human antigen. The Frizzled 9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Frizzled 9 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1G9G4 | |
Unconjugated | |
20 μL | |
8326 | |
IgG1 |