missing translation for 'onlineSavingsMsg'
Learn More
Furo[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 15487337

Furo[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15487337
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15487337

Brand: Thermo Scientific Alfa Aesar H34400.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112372-14-2
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD11588128
InChI Key VHRCHODSRLMXFM-UHFFFAOYSA-N
Synonym furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
PubChem CID 13803071
IUPAC Name furo[3,2-b]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CC2=NC=CC=C2O1

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 163.13
Percent Purity 97%
Chemical Name or Material Furo[3,2-b]pyridine-2-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10181314 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberH34400.03,H34400.MD,H34400.06
Lot Number10181314 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34400.03,H34400.MD,H34400.06
Lot Number10170074 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH34400.03,H34400.MD,H34400.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Furo[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.