missing translation for 'onlineSavingsMsg'
Learn More

GABA-A R gamma 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP3-17492-100UL

 View more versions of this product

Product Code. 18305312

  • 5775.00 SEK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GABA-A R gamma 1 Polyclonal antibody specifically detects GABA-A R gamma 1 in Human samples. It is validated for Immunofluorescence
Specifications

Specifications

GABA-A R gamma 1
Polyclonal
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
DKFZp686H2042, GABA(A) receptor subunit gamma-1, gamma-1 polypeptide, gamma-aminobutyric acid (GABA) A receptor, gamma 1, gamma-aminobutyric acid receptor subunit gamma-1, MGC33838
This antibody was developed against Recombinant Protein corresponding to amino acids: NKASMTPGLHPGSTLIPMNNISVPQEDDYGYQCLEGKDC
100 μg
Stem Cell Markers
2565
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunofluorescence
Unconjugated
PBS, pH 7.2, 40% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.