Learn More
ROBU VitraPOR™ Gas Distribution Tube, Type B
Gas Distribution Tube with lateral bowl.
Brand: ROBU VitraPOR™ 19221
Additional Details : Weight : 0.18050kg
Description
- For fine dispersion of gases in liquids
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Type B | |
1 | |
22 mm |