Learn More
Lenz Laborglasintrumente Gas Inlet Tubes, Straight
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05080003
This item is not returnable.
View return policy
Description
- Straight air leak tubes
- Built-in length: 200 mm
- With NS cone and thin capillary (easily to draw by burner)
- Made of DURAN™ tubing
Specifications
Duran Glass | |
Cone NS 24/29 |