Learn More
3M™ Gas, Vapour and Particulate Filter
3M™ Gas, Vapour and Particulate Filters have a bayonet-style connection, and they’re designed to optimise your field of vision. Our lightweight filters offer well-balanced breathing protection for hazardous environments.
Brand: 3M™ 7000030098
Additional Details : Weight : 0.59400kg
Description
Our lightweight filters combine protection with comfort. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision. They can be clicked into place quickly and easily. Features a bayonet-style connection.Specifications
Organic vapor, mercury vapor and particulates | |
A1 HgP3R | |
Typical industrial applications for filters are laboratories and particulate applications | |
6096 |
EN14387:2004 + A1:2008, CE Approved | |
Gas, Vapor and Particulate Filter | |
3M™ 6000 and 3M™ 7907 Series Full Face Masks | |
4/Pk. |