Learn More
Lenz Laborglasintrumente Gas Washing Bottles Acc. to Drechsel with Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05510249
This item is not returnable.
View return policy
Description
- Acc. to DIN 12596, complete with head (with filter disc)
- Ground joint connection: NS 29/32
- Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
250 mL | |
Olive connections | |
NS 29/32 |
Borosilicate Glass (3.3), Duran Tubing | |
2 |