missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA4 Recombinant Rabbit Monoclonal Antibody (1M2E4), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Thermo Scientific MA538112
This item is not returnable.
View return policy
Description
Positive Samples: HepG2, SKOV3, Mouse liver, Mouse testis, Rat testis Immunogen sequence: ASGASSNSSN ATTSSSEEMR PIKTEPGLSS HYGHSSSVSQ TFSVSAMSGH GPSIHPVLSA LKLSPQGYAS PVSQSPQTSS KQDSWNSLVL ADSHGDIITA
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.Specifications
GATA4 | |
Recombinant Monoclonal | |
0.55 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P46152, Q08369 | |
GATA4 | |
A synthetic peptide corresponding to a sequence within amino acids 343-442 of human GATA4 (NP_0020432) | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1M2E4 | |
Unconjugated | |
GATA4 | |
ASD2; DNA-binding protein GATA-GT2; GAT4; GATA binding protein 4; GATA4; Gata-4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; TACHD; TOF; Transcription factor GATA-4; VSD1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14463, 54254 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction