missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCFC2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
4460.00 SEK
Specifications
Antigen | GCFC2 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GCFC2 Polyclonal specifically detects GCFC2 in Human samples. It is validated for Western Blot.Specifications
GCFC2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
C2orf3, chromosome 2 open reading frame 3, DNABF, GC bindng factor, GCF, GCFGC binding factor, GC-rich sequence DNA-binding factor, GC-rich sequence DNA-binding factor 2, TCF9, TCF-9, Transcription factor 9, transcription factor 9 (binds GC-rich sequences) | |
The immunogen is a synthetic peptide directed towards the middle region of human TCF9 (NP_003194). Peptide sequence QKQKKVFEEVQDDFCNIQNILLKFQQWREKFPDSYYEAFISLCIPKLLNP | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
6936 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title