missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Gel Casting Gate
With integral silicone seals to effectively seal tray.
1520.00 SEK - 1590.00 SEK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10134782
|
Fisherbrand™
HU25-CG |
Maxi Plus Standard | N/A | ||||||
10305212
|
Fisherbrand™
HU20-CG |
Maxi Standard | N/A | ||||||
Description
The Maxi-Plus and Maxi-Plus cooled offer a 624 sample capacity when using 12 x 52 well, multichannel pipettor compatible combs. One gel can be used to screen DNA and RNA samples from 6 x 96 well gates with rubber gaskets to provide a leak proof seal. High resolution is retained on long runs by the use of the buffer re-circulation ports and the high buffer volume reserve. Again, the cooled version offers the researcher the chance to increase resolutions without sacrificing time. All combs are multichannel pipettor compatible.- Gel dimensions: 250mm x 300mm (w x l)
- Buffer volume: 3,000mL
- Maximum number of samples: 624
- Removable gel casting tray
- Buffer recirculation ports
Specifications
Gel Casting Gate | |
Integral Silicone Seals |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title