Learn More
UVP Gel documentation system gel tray stainless steel for Firstlight illuminators for gels up to 250mm x 260mm UVP
Brand:  UVP 38-0296-03
Product Code. 12883908
This item is not returnable. View return policy
Description
Description
The unique design of the Firstlight Illuminator ensures extremely uniform 302nm emission, which is critical for accurate quantitative analysis. Through use of a high density lighting system and a tuned phosphor coating, uniformity of <5% CV (Coefficient of Variance) across the surface has been achieved. Uniform illumination is critical for quantitative analysis of 2D gels particularly. It also maintains the integrity of data by making the reliance on software uniformity correction redundant.Dimensions 279 x 356 x 143mm