Learn More
Thermo Scientific™ GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 Cells
GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells contain the human calcium-sensing receptor (CaSR), (Accession No. NM_000388) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1833
Description
GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells contain the human calcium-sensing receptor (CaSR), (Accession No. NM_000388) stably integrated into the GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 (Cat. No. K1536) cells contain a beta-lactamase reporter gene under control of the NFAT Response Element
The GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Spermine. In addition, GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
NM_000388 | |
Dividing Cells | |
Fluorescence | |
CaSR | |
CaSR-Gqo5-NFAT-bla CHO-K1 cells, CaSR-Gqo5-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
End Point | |
CHO-K1 Cells | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells • GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ CaSR-Gqo5-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
FRET | |
G-Protein Coupled Receptor (GPCR) | |
Single Cell Clone Origin | |
8 x 106 Cells | |
CASR | |
CHO-K1 | |
GPCR |
For Research Use Only. Not for use in diagnostic procedures.