Learn More
Thermo Scientific™ GeneBLAzer™ SCTR-CRE-bla CHO-K1 Cells
GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells contain the human Secretin Receptor (SCTR) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
Brand: Thermo Scientific™ K1805
Description
GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells contain the human Secretin Receptor (SCTR) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
The GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of Secretin. In addition, GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
NM_002980.2 | |
Dividing Cells | |
Fluorescence | |
SCTR | |
Pool of Stable Cells | |
8 x 106 Cells | |
BLA (Beta-Lactamase) | |
Dividing Cells | |
GeneBLAzer™ |
Cell-based beta lactamase reporter gene | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells • GeneBLAzer™ SCTR-CRE-bla CHO-K1 • Certificate of Analysis The GeneBLAzer™ SCTR-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
FRET | |
SCTR-CRE-bla CHO-K1 cells, SCTR-CRE-bla CHO-K1, Certificate of Analysis | |
GeneBLAzer™ | |
End Point | |
SCTR | |
CHO-K1 | |
GPCR |