missing translation for 'onlineSavingsMsg'
Learn More
Learn More
General Purpose Reference Electrode, Hanna Instruments™
Click to view available options
Reference: Ag/AgCl
Specifications
Specifications
Description | General Purpose Reference Electrode |
For Use With (Application) | General purpose, titrations |
Chemical Name or Material | Reference electrode |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction