missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP-ATF2 (19-96)
GFP-ATF2 (19-96)
Brand: Invitrogen PV4445
This item is not returnable.
View return policy
Description
A truncated version (residues 19-96) of ATF2 has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a substrate for the JNK and p38 families of kinases.For use in combination with the LanthaScreen™ Tb-anti-ATF2 [pThr71] antibody (PV4451) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Biochemical terbium phospho-specific activity | |
Fluorescence | |
20 nmol | |
Protein-Based Substrate | |
Serine/Threonine Kinases | |
478000 Da | |
MAPK13, MAPK8, MAPK9, MAPK14, MAPK3, MAPK1, MAPK11, MAPK12, MAPK10 |
GFP-ATF2 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1mM DTT. Store at -80°C. | |
Liquid | |
ATF2 (19-96) | |
Kinase Substrate | |
GFP (EmGFP) | |
Dry Ice | |
MAPK1, MAPK3, MAPK8, MAPK9, MAPK10, MAPK11, MAPK12, MAPK13, MAPK14 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction