missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GHR Rabbit anti-Human, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20713-1-AP-150UL
This item is not returnable.
View return policy
Beschreibung
may play a role in chondrocyte differentiation and positive regulation of longitudinal bond growth and.Spezifikation
GHR | |
Polyclonal | |
Unconjugated | |
GHR | |
GH binding protein, GH receptor, GHBP, GHR, growth hormone receptor, Serum binding protein, Somatotropin receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2690, 397488 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P10912, P19756 | |
GHR | |
Peptide | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG |