Learn More
Gibco™ Fetal Bovine Serum, certified, United States
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
Brand: Gibco™ 16000069
Additional Details : Weight : 0.30000kg
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
Gibco Sera: Trusted Quality for Consistent Performance
Sera Category: Premium (Performance Plus)
- Use for a broad range of cell types, especially sensitive cell lines
- Our most characterized sera with the lowest endotoxin and hemoglobin levels
- Endotoxin level: ≤5 EU/mL
- Hemoglobin level: ≤15 mg/dL
- Origin: United States
Gibco Serum Delivers
- ISO 13485 certified, processed in FDA registered facilities
- Triple filtered at 0.1 μm
- Gibco bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
Triple-filtered, 0.1 μm | |
Sustainable packaging | |
Certified |
Fetal | |
United States | |
Bottle | |
1000 mL | |
Sterile | |
≤5 EU/mL | |
≤15 mg/dL | |
Frozen |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.