missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CCL13 (MCP-4) Recombinant Protein
Human CCL13 (MCP-4) Recombinant Protein
Brand: Gibco™ PHC1154
This item is not returnable.
View return policy
Description
Human CCL13 (MCP-4) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
MCP-4 displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils, but not neutrophils. This chemokine plays a role in accumulation of leukocytes during inflammation. It may also be involved in the recruitment of monocytes into the arterial wall during artherosclerosis. It is a member of the C-C subfamily of chemokines which are characterized by having two adjacent cysteine residues.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
8.6 kDa | |
20 μg | |
75 aa | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Chemokines & Receptors | |
Lyophilized | |
Signal Transduction, Immunology | |
98 % |
6357 | |
Sequential Chromatography | |
MCP-4 | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
MCP⁄MCAF Chemokines | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.