Learn More
Gilson™ Tip Empty Rack For Pipet Tips
Use with loose Gilson Diamond tips. Gilson™ Tip Empty Rack For Pipette Tips are color coded for ease of use.
Brand: Gilson™ F171402
242.25 SEK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Fully autoclavable
- Made of polypropylene
Specifications
Empty Rack | |
Blue/Green | |
Polypropylene | |
Yes, Interlocking | |
Diamond Tips | |
3.66 in. | |
4.88 in. |
Autoclavable | |
Pipette Tips (20 to 300 μL) | |
Rectangular | |
Non-sterile | |
9.3 cm | |
5.28 in. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
7Hours
11Minutes
36Seconds
19
Product Content Correction
Product Title
Gilson™ Tip Empty Rack For Pipet Tips > Tip Empty Rack For Pipet Tips; Holds: 20 to 300 μL D300 Pipet Tips
Spot an opportunity for improvement?Share a Content Correction