missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GIP Monoclonal Antibody (4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen ABS0210402
This item is not returnable.
View return policy
Description
ABS 021-04-02 detects GIP in human samples. ABS 021-04-02 has been successfully used in ELISA and immunohistochemistry procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
This gene encodes an incretin hormone and belongs to the glucagon superfamily. The encoded protein is important in maintaining glucose homeostasis as it is a potent stimulator of insulin secretion from pancreatic beta-cells following food ingestion and nutrient absorption. This gene stimulates insulin secretion via its G protein-coupled receptor activation of adenylyl cyclase and other signal transduction pathways. It is a relatively poor inhibitor of gastric acid secretion.
Specifications
GIP | |
Monoclonal | |
1.02 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P09681 | |
Gip | |
Synthetic human gastric inhibitory polypeptide. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
4 | |
Unconjugated | |
Gip | |
gastric inhibitory polypeptide; Gip; Glucose-dependent insulinotropic peptide; glucose-dependent insulinotropic polypeptide; Gludins; incretin hormone; Rabbit Anti GIP; RATGLUDINS | |
Mouse | |
Protein A | |
RUO | |
2695 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
GIP Monoclonal Antibody (4), Invitrogen™ > 400 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction