Learn More
Cytiva Glass Plate Kit
Used when spot picking lab-cast gels and when background fluorescence is not concerned
Brand: Cytiva 80-6475-39
Includes:
- 1 Glass plate (27.6 x 21.7cm x 3mm)
- 1 Glass plate (27.6 x 21.1cm x 3mm)
- 2 Vinyl spacer (10mm x 1mm)
Description
Specifications
Glass Plate Kit | |
1 Glass plate (27.6 x 21.7cm x 3mm), 1 Glass plate (27.6 x 21.1cm x 3mm), 2 Vinyl spacer (10 x 1mm) | |
Complete packsize |