Learn More
Hirschmann™ Hollow Glass Stoppers
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
59.80 SEK - 698.27 SEK
Specifications
Material | Glass |
---|---|
Product Type | Hollow Stopper |
For Use With (Equipment) | For tight sealing of standard ground vessels |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10014001
|
Hirschmann™
8900390 |
NS 34/35 |
190.52 SEK
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
10678971
|
Hirschmann™
8900393 |
NS 45/40 |
269.00 SEK
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
|||||
30090416
|
Hirschmann™
|
NS 7/16 |
587.71 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30090410
|
Hirschmann™
|
NS 10/19 |
587.71 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10556172
|
Hirschmann™
8900360_OLD |
59.80 SEK
Each |
In Stock
Log in to see stock. |
||||||
30090418
|
Hirschmann™
8900365 |
NS 12/21 |
587.71 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30090396
|
Hirschmann™
8900375 |
NS 19/26 |
698.27 SEK
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10238111
|
Hirschmann™
8900380 |
NS 24/29 |
88.75 SEK
Each |
Estimated Shipment: 23-10-2024
Log in to see stock. |
|||||
10524781
|
Hirschmann™
8900386 |
NS 29/32 |
98.25 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Acc. to DIN 12 252
- Available to fit a range of sizes
Specifications
Glass | |
Hollow Stopper | |
For tight sealing of standard ground vessels |