Learn More
Glassy carbon rod, 1mm (0.04in) dia, type 1, Thermo Scientific Chemicals
Widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.
2319.42 SEK - 6934.58 SEK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384327
|
Thermo Scientific Alfa Aesar
038009.DJ |
50 mm |
2319.42 SEK
50mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11304337
|
Thermo Scientific Alfa Aesar
038009.DM |
100 mm |
4106.31 SEK
100mm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15421477
|
Thermo Scientific Alfa Aesar
038009.DR |
200 mm |
6934.58 SEK
200mm |
Estimated Shipment: 30-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Glassy carbon rod is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.