Learn More
Thermo Scientific Chemicals Glove Bag Clamp Product Code.: 15491837
Thermo Scientific Chemicals Glove Bag Clamp
Product Code. 15491837
1 Ea.
Click to view available options
Quantity:
1 Ea.
Product Code. 15491837
Brand: Thermo Scientific Chemicals 042909.KF
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Quick-closing clamp
  • For use with 17 x 17 in. polyethylene glove bags
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Polyethylene Glove Bags (17 x 17 in.)
Quantity 1 Ea.
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Glove Bag Clamp >