missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose 6 Phosphate Dehydrogenase Rabbit anti-Human, Clone: 6V8O9, Novus Biologicals™
Rabbit Monoclonal Antibody
1905.00 SEK - 4805.00 SEK
Specifications
Antigen | Glucose 6 Phosphate Dehydrogenase |
---|---|
Clone | 6V8O9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321495
|
Bio-Techne
NBP3-15359-20UL |
20 μg |
1905.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18335014
|
Bio-Techne
NBP3-15359-100UL |
100 μg |
4805.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western BlotSpecifications
Glucose 6 Phosphate Dehydrogenase | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Golgi Apparatus Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2539 | |
IgG | |
Affinity purified |
6V8O9 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human Glucose 6 Phosphate Dehydrogenase (P11413). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title