missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Reduced Crystalline MP Biomedicals
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
100 g
Specifications
Specifications
Color | White |
Assay Percent Range | >98% |
Quantity | 10 g |
Formula Weight | 307.3 |
Physical Form | Powder |
Chemical Name or Material | Glutathione, Reduced |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction