missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Honeywell™ Riedel-de Haen™
BIOPHARMA

Product Code. 15601480
Click to view available options
Quantity:
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E422, anhydrous, 99.0-101.0% (alkalimetric)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C
Boiling Point 290°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Ignition Point 698°F
Packaging Poly Bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index n20/D 1.474(lit.)
Quantity 4 x 5 L
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1mmHg (20°C)
Percent Purity 99.0 to 101.0% (calc. on H2O free substance)
Grade puriss.
Physical Form Liquid
Chemical Name or Material Glycerol, Meets analytical specification of Ph. Eur. BP, USP, FCC.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'