missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl phenyl ether, 99%, Thermo Scientific Chemicals

666.07 SEK - 1893.28 SEK

Chemical Identifiers

CAS 122-60-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00005133,MFCD31699959
InChI Key FQYUMYWMJTYZTK-UHFFFAOYNA-N
Synonym glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
PubChem CID 31217
ChEBI CHEBI:82367
IUPAC Name 2-(phenoxymethyl)oxirane
SMILES C(OC1=CC=CC=C1)C1CO1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10606342
SDS View Documents
Thermo Scientific Acros
135350500
50 g Glass bottle
666.07 SEK
50g
Please to purchase this item. Need a web account? Register with us today!
10357210
SDS View Documents
Thermo Scientific Acros
135352500
250 g Glass bottle
1893.28 SEK
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

122-60-1
150.18
FQYUMYWMJTYZTK-UHFFFAOYNA-N
31217
2-(phenoxymethyl)oxirane
C9H10O2
MFCD00005133,MFCD31699959
glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
CHEBI:82367
C(OC1=CC=CC=C1)C1CO1

Specifications

122-60-1
Colorless to Yellow
245.0°C
Authentic
C9H10O2
MFCD00005133,MFCD31699959
glycidyl phenyl ether, phenyl glycidyl ether, 2-phenoxymethyl oxirane, 1,2-epoxy-3-phenoxypropane, oxirane, phenoxymethyl, phenylglycidyl ether, 2,3-epoxypropyl phenyl ether, phenoxypropene oxide, phenylglycydyl ether, ether, phenylglycidyl
FQYUMYWMJTYZTK-UHFFFAOYNA-N
2-(phenoxymethyl)oxirane
31217
7 mPa.s (20°C)
99%
Glycidyl phenyl ether, 99%
3.5°C
1.1000g/mL
115°C
98.5% min. (GC)
1.5295 to 1.5315
1.1
Solubility in water: 2.4g/L(20°C). Other solubilities: miscible with ethanol
C(OC1=CC=CC=C1)C1CO1
150.18
CHEBI:82367
150.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-557-2

missing translation for 'rtecsNumber' : TZ3675000

missing translation for 'tsca' : TSCA

Product Identifier
  • Glycidyl phenyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.