missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GlycoLink™ Micro Immobilization Kit

Immobilize polyclonal antibodies or other glycoproteins, via periodate-oxidized sugars, onto hydrazide-activated polyacrylamide beads for AP, IP or pull-down assays.

Brand:  Thermo Scientific™ 88942

Product Code. 11638843

  • 4475.00 SEK / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ GlycoLink Immobilization Kits provide affinity columns and reagents to immobilize glycoproteins through oxidized sugar groups for use in affinity-purification applications or studies of protein-protein interactions.

The GlycoLink Kits use hydrazide-activated resin and a special catalyst to prepare affinity columns with polyclonal antibodies, which typically have abundant carbohydrates (glycosylation) on their Fc portions. Any glycosylated proteins (even monoclonal antibodies) that contain periodate-oxidizable carbohydrate groups can be immobilized with these kits to make reusable affinity columns. Antibodies immobilized by this hydrazide immobilization chemistry have unobstructed antigen-binding sites and optimal purification capability. Prepared columns with stable glycoproteins or antibodies can be regenerated and reused at least five times for affinity-purification without significant loss in binding capacity.

Highlights:

  • High-capacity – immobilize 1 to 10mg of oxidized antibody or other glycoprotein per milliliter of resin

  • Fast and efficient – immobilize in as few as 30 minutes; achieve at least 90% coupling of most glycoproteins in less than 4 hours (efficiency depends on the amount and type of glycosylation)

  • Specific immobilization – the hydrazide-activated UltraLink Resin conjugates only to purified glycoproteins whose sugar (e.g., sialic acid) groups have been gently oxidized with periodate

  • Stable attachment – resonance structure of the hydrazone bonds are sufficiently stable for multiple affinity purifications with one batch of prepared resin; no stabilization step is required

  • Preserves antibody function – immobilizes IgG via the Fc region, thereby keeping both antigen binding sites available for capturing target

  • Convenient sizes – choose from two complete 10-column kits: one with 1mL columns and one with 0.1mL columns

UltraLink Hydrazide Resin:

  • Ligand: hydrazide
  • Support: Thermo Scientific UltraLink Biosupport
  • Spacer arm: 22 atoms
  • Loading: activated with at least 15μmol hydrazide per milliliter of settled resin
  • Format: 50% resin slurry in 0.02% sodium azide

Includes:

Kits contain GlycoLink Columns; coupling buffer, coupling catalyst and wash buffer; oxidizing reagent; desalting columns, PBS buffer pack; and column accessories

Recommended for:

Purify antigens using covalently coupled antibodies; Determine binding partners for a glycoprotein of interest; Perform large-scale affinity purifications

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Immobilizing 50 to 500 μg of GlycoProtein per Column
Sufficient For: Immobilizing 50 to 500 μg of glycoprotein per column
• GlycoLink Columns, 0.1 mL, 10 columns
• GlycoLink Coupling Buffer, 100 mL
• Aniline, 0.2 mL
• Wash Buffer, 60 mL
• Sodium meta-periodate, 0.5 g
• Zeba Spin Desalting Columns, 0.5 mL, 10 columns
• PBS Pack (makes 500mL), 1 pack
• Column Accessories, 10 ea

Store at 4°C.
10 Columns
Hydrazide
Kit, Microcentrifuge, Spin Column
GlycoLink Micro Immobilization Kit
GlycoLink™
Immobilization Kit
10 Columns
Affinity, UltraLink™ Resin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.