Learn More
Glyoxal sodium bisulfite hydrate, 98%, Thermo Scientific™
Product Code. 15567029
Click to view available options
Quantity:
25 g
250 g
1 kg
Chemical Identifiers
CAS 332360-05-1
Molecular Formula C2H4Na2O8S2
Molecular Weight (g/mol) 266.15
MDL Number MFCD0149541
InChI Key BXUKAXFDABMVND-UHFFFAOYNA-L
Synonym glyoxal-sodium bisulfite monohydrate, glyoxal sodium bisulfite addition compound hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt hydrate, 1,2-dihydroxy-1,2-ethanedisulfonic acid disodium salt, c2h4o8s2.2na.h2o, glyoxal-sodium bisulfite monohydrate mi, glyoxal bis sodium hydrogen sulfite adduct hydrate, glyoxal sodium bisulfite addition compound hydrate,98, disodium 1,2-dihydroxy-1,2-ethanedisulfonate monohydrate, disodium 1,2-dihydroxyethane-1,2-disulfonate hydrate
PubChem CID 16211366
SMILES [Na+].[Na+].OC(C(O)S([O-])(=O)=O)S([O-])(=O)=O
Specifications
Melting Point 193.0°C to 195.0°C
Quantity 1 kg
Assay Percent Range 97.5% min
Linear Formula [-CH(OH)SO3Na]2·xH2O
Merck Index 14,451
Solubility Information Solubility in water: freely soluble. Other solubilities: practically insoluble in alcohol
IUPAC Name disodium 1,2-dihydroxyethane-1,2-disulfonate
Formula Weight 266.16
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 10% max. (120°C, 2 hrs)
Chemical Name or Material Glyoxal sodium bisulfite hydrate