Learn More
Goat anti-Chicken IgG whole molecule, FITC, Polyclonal, Secondary Antibody, Abnova™
Brand:  Abnova PAB10499.2mg
Product Code. 16242840
  • 3335.00 SEK / 2mg
Explore more special offers
This item is not returnable. View return policy
Specifications
Goat anti-Chicken IgG whole molecule
Polyclonal
Goat anti-chicken IgG recognizes chicken IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.
Lyophilized
Chicken IgG whole molecule
2 mg
Chicken
Lyophilized
Flow Cytometry, Immunofluorescence
FITC
The optimal working dilution should be determined by the end user.
Goat
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Secondary
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.