missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, BODIPY™ FL, Invitrogen™

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen B2752

Product Code. 10544583

  • 1526.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.

This item is not returnable. View return policy

Description

Description

Storage and reconstitution: reconstitute using 0.5 mL deionized water. The product can be stored up to 2 weeks at 2-8oC. For longer storage, divide into single-use aliquots and freeze at or below -20oC. Frozen aliquots are stable for at least 6 months.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG (H+L) Cross-Adsorbed
Polyclonal
PBS with 1.5% BSA and 0.01% thimerosal; pH 7.5
Gamma Immunoglobins Heavy and Light chains
1 mg
Secondary
-20° C, store in dark
IgG
Immunocytochemistry
BODIPY FL
Goat
purified
RUO
Mouse
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers