missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgG (H+L) Secondary Antibody, Janelia Fluor 549 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72716JF549
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
IgG (H+L) | |
Polyclonal | |
50mM Sodium Borate | |
Rat IgG whole molecule | |
0.5 mL | |
Adaptive Immunity, Immunology | |
Rat | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Janelia Fluor 549 | |
Goat | |
Multi-step | |
RUO | |
Secondary | |
Store at 4°C in the dark. |