missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
4460.00 SEK
Specifications
Antigen | GOLGA3 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GOLGA3 Polyclonal specifically detects GOLGA3 in Human samples. It is validated for Western Blot.Specifications
GOLGA3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
GCP170Golgi peripheral membrane protein, golgi autoantigen, golgin subfamily a, 3, Golgi complex-associated protein of 170 kD, Golgi complex-associated protein of 170 kDa, Golgi membrane associated protein, golgin A3, Golgin subfamily A member 3, Golgin-160, golgin-165, male enhanced antigen-2, MEA-2, SY2/SY10 protein | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Human GOLGA3. Peptide sequence VGSLGLPAHPREKKTSKSSKIRSLADYRTEDSNAGNSGGNVPAPDSTKGS | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
2802 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title