Learn More
Thermo Scientific™ Tango™ GPR1-bla U2OS Cells
Tango™ GPR1-bla U2OS cells contain the human G protein-coupled receptor 1 (GPR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1839
Additional Details : Weight : 0.01000kg
Description
Tango™ GPR1-bla U2OS cells contain the human G protein-coupled receptor 1 (GPR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ GPR1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Chemerin (Figure 1). In addition, Tango™ GPR1-bla U2OS cells have been tested for assay performance under variable conditions.
Specifications
NM_005279.3 | |
Dividing Cells | |
Native Tail | |
FRET | |
G-Protein Coupled Receptor (GPCR) | |
Single Cell Clone Origin | |
8 x 106 Cells | |
GPR1 | |
U2OS | |
GPCR |
Cell-based beta-arrestin recruitment | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ GPR1-UAS-bla CHO-K1 cells • Tango™ GPR1-UAS-bla CHO-K1 Assay Protocol • Certificate of Analysis The Tango™ GPR1-UAS-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
GPR1 | |
GPR1-UAS-bla CHO-K1 cells, GPR1-UAS-bla CHO-K1 Assay Protocol, Certificate of Analysis | |
Tango™ | |
End Point | |
Dividing Cells | |
Tango™ |
For Research Use Only. Not for use in diagnostic procedures.