Learn More
GPR101 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551510
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RMEAKDGSLK AKEGSTGTSE SSVEARGSEE VRESSTVASD GSMEGKEGST KVEENSMKAD KGRTEVNQCS IDLGEDDMEF GEDDINFSED DVEAVNIPES LPPSRRNSNS NPPLPRCYQC KAA Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 57%.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. Expression of GPR101 has been documented by Northern analysis in human hypothalamus, caudate, and putamen but not in thalamus, hippocampus, frontal cortex, or pons.Specifications
GPR101 | |
Polyclonal | |
Unconjugated | |
GPR101 | |
G protein-coupled receptor 101; G protein-coupled receptor GPR101; Gm365; GPCR6; Gpr101; PAGH2; Probable G-protein coupled receptor 101; RGD1564196 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83550 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96P66 | |
GPR101 | |
Recombinant protein corresponding to Human GPR101 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only