missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR114 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521701
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the adhesion family of G-protein coupled receptors. Members of this family are characterized by long N-termini and multiple functional domains. They may play a role in the immune system as well as in the central nervous system. Alternative splicing results in multiple transcript variants.Specifications
GPR114 | |
Polyclonal | |
Unconjugated | |
ADGRG5 | |
ADGRG5; adhesion G protein-coupled receptor G5; adhesion G-protein coupled receptor G5; G protein-coupled receptor 114; G protein-coupled receptor PGR27; GPR114; G-protein coupled receptor 114; G-protein coupled receptor PGR27; PGR27; probable G-protein coupled receptor 114; UNQ2524/PRO6017 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221188 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8IZF4 | |
ADGRG5 | |
Recombinant fragment corresponding to a region within amino acids 2 and 283 of GPR114 (Uniprot ID#Q8IZF4) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |