missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPX8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1680.00 SEK - 3415.00 SEK
Specifications
Antigen | GPX8 |
---|---|
Dilution | Western Blot 1:1000 - 1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321374
|
Bio-Techne
NBP3-16011-20UL |
20 μg |
1680.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18337395
|
Bio-Techne
NBP3-16011-100UL |
100 μg |
3415.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPX8 Polyclonal antibody specifically detects GPX8 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
GPX8 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 1.11.1, EC 1.11.1.9, EPLA847, glutathione peroxidase 8 (putative), GPx-8, GSHPx-8, probable glutathione peroxidase 8, UNQ847 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 41-209 of human GPX8 (NP_001008398.2). KPKINSFYAFEVKDAKGRTVSLEKYKGKVSLVVNVASDCQLTDRNYLGLKELHKEFGPSHFSVLAFPCNQFGESEPRPSKEVESFARKNYGVTFPIFHKIKILGSEGEPAFRFLVDSSKKEPRWNFWKYLVNPEGQVVKFWKPEEPIEVIRPDIAALVRQVIIKKKEDL | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot 1:1000 - 1:2000 | |
Polyclonal | |
Purified | |
RUO | |
PBS, 50% glycerol, pH7.3 | |
493869 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title